Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylhydrazine, 97%

Catalog No. p-3737206
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296681000 100 g Glass bottle
AC296685000 500 g Glass bottle
AC296680025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC296681000 Supplier Thermo Scientific Chemicals Supplier No. 296681000
Only null left
Add to Cart

CAS: 100-63-0 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-63-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00007573
InChI Key HKOOXMFOFWEVGF-UHFFFAOYSA-N
Synonym hydrazinobenzene, hydrazine, phenyl, phenylhydrazin, monophenylhydrazine, phenyl hydrazine, 1-phenylhydrazine, hydrazine-benzene, fenilidrazina, fenylhydrazine, fenylhydrazine dutch
PubChem CID 7516
ChEBI CHEBI:27924
IUPAC Name phenylhydrazine
SMILES NNC1=CC=CC=C1

Specifications

Melting Point 19.0°C
Color Beige or Blue to White
Density 1.0900g/mL
Boiling Point 238.0°C to 241.0°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.6040 to 1.6090
Linear Formula C6H5NHNH2
Quantity 100 g
Beilstein 15,II,44
Fieser 01,838; 02,322; 06,457; 07,284
Merck Index 15,7404
Specific Gravity 1.09
Solubility Information Solubility in water: 145g/L (20°C). Other solubilities: soluble in diluted acids,miscible with alcohol,ether,chloroform and,benzene,sparingly soluble in petroleum ether
Formula Weight 108.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Phenylhydrazine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
Suspected of causing genetic defects.
May cause cancer.
Toxic if inhaled.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 202-873-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.