Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylhydrazine Hydrochloride 98.5+%, TCI America™
SDP

Catalog No. P0184100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0184100G 100 g
P018425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0184100G Supplier TCI America Supplier No. P0184100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59-88-1
Molecular Formula C6H9ClN2
Molecular Weight (g/mol) 144.60
MDL Number MFCD00012924
InChI Key JOVOSQBPPZZESK-UHFFFAOYSA-N
Synonym phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
PubChem CID 60962
IUPAC Name hydrogen phenylhydrazine chloride
SMILES [H+].[Cl-].NNC1=CC=CC=C1

Specifications

Melting Point 254°C
Color White-Yellow
Quantity 100 g
UN Number 2811
Formula Weight 144.60
Percent Purity ≥98.5% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylhydrazine Hydrochloride
EINECSNumber (3)-0470
RTECSNumber MV9000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.