Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylhydrazine Oxalate 98.0+%, TCI America™
SDP

Catalog No. P018525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P018525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P018525G Supplier TCI America Supplier No. P018525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37887-33-5
Molecular Formula C6H8N2
MDL Number MFCD00060204

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 306.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylhydrazine Oxalate
EINECSNumber (3)-0470
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.