Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylhydrazine Sulfate 98.0+%, TCI America™
SDP

Catalog No. P018625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P018625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P018625G Supplier TCI America Supplier No. P018625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52033-74-6
Molecular Formula C6H8N2
MDL Number MFCD00070608

Specifications

Quantity 25 g
UN Number 2811
Formula Weight 314.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylhydrazine Sulfate
EINECSNumber (3)-0470
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.