Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylhydroquinone 98.0+%, TCI America™
SDP

Catalog No. D05845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05845G 5 g
D058425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D05845G Supplier TCI America Supplier No. D05845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1079-21-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00002342
InChI Key XCZKKZXWDBOGPA-UHFFFAOYSA-N
Synonym phenylhydroquinone, 2-phenylhydroquinone, 2,5-dihydroxybiphenyl, 1,1'-biphenyl-2,5-diol, biphenyl-2,5-diol, hydroquinone, phenyl, 1,4-benzenediol, phenyl, 2,5-biphenyldiol, o-phenylhydroquinone
PubChem CID 14116
IUPAC Name [1,1'-biphenyl]-2,5-diol
SMILES OC1=CC=C(O)C(=C1)C1=CC=CC=C1

Specifications

Melting Point 101°C
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenylhydroquinone
EINECSNumber (4)-1336
RTECSNumber DV4550000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.