Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylmagnesium bromide, 3M in 2-MeTHF

Catalog No. aah54729ae
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54729AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH54729AE Supplier Thermo Scientific Chemicals Supplier No. H54729AE
Only null left
Add to Cart

CAS: 100-58-3 | C6H5BrMg | 181.32 g/mol

Phenyl magnesium bromide is the suitable reagent used for the synthesis of end-functionalized regioregular poly(3-alkylthiophene)s. It may be used for synthesis of  1,3,3-trimethyl-6-phenyl-2-oxabicyclo[2.2.2]octan-6-ol, 6-benzyl-1,3,3-trimethyl-2-oxabicyclo[2.2.2]octan-6-ol, (3-(2-Dithiobenzoatepropionyl)propyl)dimethylmethoxysilane, reversible addition-fragmentation chain transfer polymerization (RAFT)-silane agent, series of o-substituted benzophenones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl magnesium bromide is the suitable reagent used for the synthesis of end-functionalized regioregular poly(3-alkylthiophene)s. It may be used for synthesis of  1,3,3-trimethyl-6-phenyl-2-oxabicyclo[2.2.2]octan-6-ol, 6-benzyl-1,
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-58-3
Molecular Formula C6H5BrMg
Molecular Weight (g/mol) 181.32
MDL Number MFCD00000038
InChI Key ANRQGKOBLBYXFM-UHFFFAOYSA-M
Synonym phenylmagnesium bromide, bromophenylmagnesium, magnesium, bromophenyl, phmgbr, phenylmagnesiumbromide, bromo phenyl magnesium, phenyl magnesium bromide, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide, grignard reagent
PubChem CID 66852
IUPAC Name magnesium;benzene;bromide
SMILES Br[Mg]C1=CC=CC=C1

Specifications

Linear Formula C6H5MgBr
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 181.31
Concentration or Composition (by Analyte or Components) 3M in 2-MeTHF
Chemical Name or Material Phenylmagnesium bromide
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.