Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenylmagnesium Bromide Solution

Catalog No. 331376800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
100 mL
4 x 25 mL
800 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-137-6800ML 800 mL
33-137-6100ML 100 mL
33-137-625ML 4 x 25 mL
33-137-650ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 33-137-6800ML Supplier Sigma Aldrich Supplier No. 331376800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phenylmagnesium bromide (PhMgBr) solution is an organomagnesium reagent mainly used in Grignard reaction, which finds application as the key step in the synthesis of (S)-2(diphenylmethyl)pyrrolidine, a chiral solvating agent for the NMR analysis. PhMgBr can also undergo palladium-catalyzed Grignard coupling with alkyl chlorides to form the cross-coupled product in good yield.Storage below 25°C may cause crystallization. War mgently to redissolve solid.

Specifications

CAS 100-58-3
Density 1.004 g/mL at 25°C
Molecular Formula C6H5BrMg
Linear Formula C6H5MgBr
Quantity 800 mL
MDL Number MFCD00000038
Synonym PhMgBr solution
Molecular Weight (g/mol) 181.31 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.