Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenylmagnesium Bromide Solution

Catalog No. 171565800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
100 mL
4 x 25 mL
800 mL
18 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-156-5800ML 800 mL
17-156-5100ML 100 mL
17-156-518L 18 L
17-156-550ML 50 mL
17-156-525ML 4 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Danger
Catalog No. 17-156-5800ML Supplier Sigma Aldrich Supplier No. 171565800ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phenylmagnesium bromide solution contains 3M phenylmagnesium bromide in diethyl ether. It can act as a strong acid and Lewis acid. It can undergo addition reaction with many unsaturated functional groups. The phenyl group can displace halide from other organic compounds. Phenylmagnesium bromide is a Grignard reagent. Reaction of +a-cyclohexanedione (dihydroresorcinol) with phenylmagnesium bromide has been investigated.Phenylmagnesium bromide was used for the synthesis of end-functionalized regioregular poly(3-alkylthiophene)s. It was also used for the monoalkylation of aliphatic primary amine to generate secondary amines by the Grignard reaction of 1-[(alkylamino) methyl] benzotriazoles. It may be used for synthesis of the following: 1,3,3-trimethyl-6-phenyl-2-oxabicyclo[2.2.2]octan-6-ol 6-benzyl-1,3,3-trimethyl-2-oxabicyclo[2.2.2]octan-6-ol (3-(2-Dithiobenzoatepropionyl)propyl)dimethylmethoxysilane, reversible addition-fragmentation chain transfer polymerization (RAFT)-silane agent series of o-substituted benzophenonesStorage below 25°C may cause formation of crystalline magnesium salts. Moving container to a war mLocation and occasional gentle swirling should redissolve the solid

Specifications

CAS 100-58-3
Density 1.134 g/mL at 25°C
Molecular Formula C6H5BrMg
Linear Formula C6H5MgBr
MDL Number MFCD00000038
Quantity 800 mL
Molecular Weight (g/mol) 181.31 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.