Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenylmagnesium Chloride Solution

Catalog No. 22444818LKL Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
8 L
18 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-444-818LKL 18 L
22-444-8800ML 800 mL
22-444-8100ML 100 mL
22-444-88L 8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 22-444-818LKL Supplier Sigma Aldrich Supplier No. 22444818LKL
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phenylmagnesium chloride (PhMgCl) solution is a common Grignard reagent used in the synthesis of (-)-phenserine and stephacidin B. It is employed in a variety of cross-coupling reactions. It can also be used as an electrolyte solution along with aluminium chloride (AlCl3) in the rechargeable magnesium batteries.Storage below 25°C may cause formation of crystalline magnesium salts. Moving container to a war mLocation and occasional gentle swirling should redissolve the solid

Specifications

CAS 100-59-4
Density ∼1.02 g/mL at 20°C, 1.042 g/mL at 25°C
Molecular Formula C6H5ClMg
Linear Formula C6H5MgCl
MDL Number MFCD00000464
Quantity 18 L
Molecular Weight (g/mol) 136.86 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.