Learn More
Medchemexpress LLC Phenylmethan-d2-ol | 21175-64-4 | MFCD00084124 | 99.0% | 110.15 | C7H6D2O | 5 MG
SDP
Catalog No. 5000280958
Encompass_Preferred
Phenylmethan-d2-ol is a deuterium-labeled benzyl alcohol used as a stable isotope tracer and reference standard in analytical chemistry, mass spectrometry, and synthetic labeling. It is characterized by molecular formula C7H6D2O, molecular weight 110.15 g/mol, and a reported chemical purity of 99.0%.
  • Deuterium-labeled benzyl alcohol for tracer and analytical use.
  • High chemical purity: 99.0%.
  • Molecular formula C7H6D2O and molecular weight 110.15 g/mol.
  • Available in small quantities suitable for analytical standards, including 5 mg packages.
  • Supplied with a datasheet and certificate of analysis for batch verification.
Catalog No. 50-002-80958 Supplier Medchemexpress LLC Supplier No. HYW450789S5MG
Phenylmethan-d2-ol is a deuterium-labeled benzyl alcohol used as a stable isotope tracer and reference standard in analytical chemistry, mass spectrometry, and synthetic labeling. It is characterized by molecular formula C7H6D2O, molecular weight 110.15 g/mol, and a reported chemical purity of 99.0%.
  • Deuterium-labeled benzyl alcohol for tracer and analytical use.
  • High chemical purity: 99.0%.
  • Molecular formula C7H6D2O and molecular weight 110.15 g/mol.
  • Available in small quantities suitable for analytical standards, including 5 mg packages.
  • Supplied with a datasheet and certificate of analysis for batch verification.