Learn More
Medchemexpress LLC Phenylmethan-d2-ol | 21175-64-4 | MFCD00084124 | 99.0% | 110.15 g/mol | C7H6D2O | 10 MG
SDP
Catalog No. 5000281727
Encompass_Preferred
Phenylmethan-d2-ol is a deuterium-labeled analogue of phenylmethanol intended for research use. It serves as an internal standard and tracer for analytical and metabolic studies, and supports quantitation by NMR, GC-MS, and LC-MS.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity for reliable analytical results.
  • Suitable for tracer and metabolic studies.
  • Supplied with datasheet, COA, and SDS for quality control.
Catalog No. 50-002-81727 Supplier Medchemexpress LLC Supplier No. HYW450789S10MG
Phenylmethan-d2-ol is a deuterium-labeled analogue of phenylmethanol intended for research use. It serves as an internal standard and tracer for analytical and metabolic studies, and supports quantitation by NMR, GC-MS, and LC-MS.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity for reliable analytical results.
  • Suitable for tracer and metabolic studies.
  • Supplied with datasheet, COA, and SDS for quality control.