Learn More
Medchemexpress LLC Phenylmethan-d2-ol | 21175-64-4 | MFCD00084124 | 99.0% | 110.15 g/mol | C7H6D2O | 100 MG
SDP
Catalog No. 5000281069
Encompass_Preferred
Phenylmethan-d2-ol is the deuterium-labeled form of phenylmethanol (benzyl alcohol) used as a stable isotope reagent in analytical and synthetic chemistry. It is provided in small laboratory pack sizes for use in tracer studies, method development, and isotopic labeling workflows.
  • Deuterium-labeled benzyl alcohol for tracer and standard applications.
  • High chemical purity and isotopic labeling (99.0%).
  • Available in small milligram pack sizes suitable for analytical workflows.
  • Suitable as an internal standard for mass spectrometry and quantitative analysis.
  • Compatible with standard organic synthesis and labeling procedures.
Catalog No. 50-002-81069 Supplier Medchemexpress LLC Supplier No. HYW450789S100MG
Phenylmethan-d2-ol is the deuterium-labeled form of phenylmethanol (benzyl alcohol) used as a stable isotope reagent in analytical and synthetic chemistry. It is provided in small laboratory pack sizes for use in tracer studies, method development, and isotopic labeling workflows.
  • Deuterium-labeled benzyl alcohol for tracer and standard applications.
  • High chemical purity and isotopic labeling (99.0%).
  • Available in small milligram pack sizes suitable for analytical workflows.
  • Suitable as an internal standard for mass spectrometry and quantitative analysis.
  • Compatible with standard organic synthesis and labeling procedures.