Learn More
Sigma Aldrich Fine Chemicals Biosciences Phenylmethanesulfonyl fluoride, 329-98-6, MFCD00007424, 1 g
Catalog No. NC0533307
Encompass
Empirical Formula (Hill Notation): C7H7FO2S, Molecular Weight: 174.19, ≥98.5% (GC), Synonym: α-Toluenesulfonyl fluoride, Benzylsulfonyl fluoride, PMSF, Phenylmethylsulfonyl fluoride. Phenylmethanesulfonyl fluoride has been used in following applications: cell fractionation. Used as a supplement in nuclear protein extraction. Inhibitor of cholesterol esterase (CE) and pseudocholinesterase (PCE). Used for the collection of blood prior to centrifugation to quantify plasma ANP levels.
Catalog No. NC0533307 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P76261G
Add to Cart
Add to Cart
Empirical Formula (Hill Notation): C7H7FO2S, Molecular Weight: 174.19, ≥98.5% (GC), Synonym: α-Toluenesulfonyl fluoride, Benzylsulfonyl fluoride, PMSF, Phenylmethylsulfonyl fluoride. Phenylmethanesulfonyl fluoride has been used in following applications: cell fractionation. Used as a supplement in nuclear protein extraction. Inhibitor of cholesterol esterase (CE) and pseudocholinesterase (PCE). Used for the collection of blood prior to centrifugation to quantify plasma ANP levels.