Learn More
Medchemexpress LLC Phenylmethyl ester | 831217-40-4 | UPCMLD00WMAL3-101 | 98.2% | 931.12 g/mol | C54H66N4O10 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12480525MG
Encompass_Preferred
MAL3-101 is a small-molecule allosteric inhibitor of HSP70 used in biochemical and cellular research to modulate HSP70 ATPase activity and disrupt Hsp40 co-chaperone interactions. It is a membrane-permeable dihydropyrimidine analog supplied as a solid powder for laboratory use.
  • Allosteric inhibitor of HSP70 ATPase activity.
  • Disrupts Hsp40 co-chaperone interaction.
  • Demonstrated activity in cellular antiproliferative studies.
  • High purity suitable for research (>98%).
  • Powder form with DMSO solubility for assay preparation.
  • Recommended storage: powder at -20°C; in solvent at -80°C for longer storage.
Catalog No. 50-002-37786