Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylmethylsulfonyl Fluoride, MP Biomedicals™

Catalog No. MP21953812 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21953812 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP21953812 Supplier MP Biomedicals, Inc Supplier No. 02195381.2
Add to Cart
Add to Cart

Irreversibly inhibits serine proteases, such as trypsin and chymotrypsin. Phenylmethylsulfonyl Fluoride, MP Biomedicals also inhibits mammalian acetyl cholinesterase (but not those from fish sources) and cysteine proteases (reversible by reduced thiols).

Administration of PMSF produces analgesia unrelated to its anticholinesterase effect, and prolongs the analgesic effect of centrally administered β-endorphin..

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329-98-6
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.189
InChI Key YBYRMVIVWMBXKQ-UHFFFAOYSA-N
Synonym phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride
PubChem CID 4784
ChEBI CHEBI:8102
IUPAC Name phenylmethanesulfonyl fluoride
SMILES C1=CC=C(C=C1)CS(=O)(=O)F

Specifications

Melting Point 89°C to 99°C
Quantity 250 mg
Solubility Information Soluble in methanol and ethanol
Formula Weight 174.2
Concentration or Composition (by Analyte or Components) Working Concentration: Recommended concentration is 15-175ug/mL.
Physical Form Crystals
Recommended Storage Store at 4°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.