Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylphosphine

Catalog No. AA3015506
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA3015506 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3015506 Supplier Thermo Scientific Chemicals Supplier No. 03015506
Only null left
Add to Cart

CAS: 638-21-1 | C6H7P | 110.10 g/mol

It is mainly used as a precursor to other organophosphorus compounds. It can function as a ligand in coordination chemistry. Phenylphosphine also have uses in polymer synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 638-21-1
Density 1.00 g/mL
Boiling Point 160°C
Odor Stench
Molecular Formula C6H7P
Quantity 5 g
MDL Number MFCD00002094
UN Number UN2845
Sensitivity Air and moisture sensitive
Synonym phenylphosphine, phosphine, phenyl, phenyl phosphine, fenylfosfin, fenylfosfin czech, 1-phenylphosphane, monophenylphosphine, phenylphosphine #, phenylphosphine 1g, phosphine, phenyl-,
Solubility Information Insoluble in water.
InChI Key RPGWZZNNEUHDAQ-UHFFFAOYSA-N
SMILES PC1=CC=CC=C1
IUPAC Name phenylphosphane
Molecular Weight (g/mol) 110.10
PubChem CID 12519
Formula Weight 110.1
Physical Form Liquid
Chemical Name or Material Phenylphosphine
Show More Show Less
Hazard Category H227-H250-H252-H315-H319-H332-H335
Hazard Statement GHS H Statement
H250-H252-H332-H315-H319-H335-H227
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P222-P235-P235+P410-P261-P264b-P271-P280-P302+P334-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packaging Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 211-325-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.