Learn More
Medchemexpress LLC Phenylphosphinic aci 250g | 1779-48-2 | 142.09 g·mol⁻¹ | C6H7O2P | 250 G
SDP
Catalog No. 5000229826
Encompass_Preferred
Phenylphosphinic acid (CAS 1779-48-2) is an organophosphorus reagent used to functionalize metal oxide surfaces and as a precursor in nanoparticle and sol-gel syntheses. It is supplied as a solid research reagent for laboratory materials modification and synthesis workflows.
  • Binds to metal oxide surfaces for modification and functionalization.
  • Suitable for nanoparticle preparation and sol-gel synthesis.
  • Available as a solid research reagent in laboratory pack sizes.
  • Reported molecular weight 142.09 g·mol⁻¹ and formula C6H7O2P.
  • High purity for research use (reported ≥99% by third-party suppliers).
Catalog No. 50-002-29826 Supplier Medchemexpress LLC Supplier No. HY116210250G
Add to Cart
Add to Cart
Phenylphosphinic acid (CAS 1779-48-2) is an organophosphorus reagent used to functionalize metal oxide surfaces and as a precursor in nanoparticle and sol-gel syntheses. It is supplied as a solid research reagent for laboratory materials modification and synthesis workflows.
  • Binds to metal oxide surfaces for modification and functionalization.
  • Suitable for nanoparticle preparation and sol-gel synthesis.
  • Available as a solid research reagent in laboratory pack sizes.
  • Reported molecular weight 142.09 g·mol⁻¹ and formula C6H7O2P.
  • High purity for research use (reported ≥99% by third-party suppliers).