Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenylphosphonic acid | 1571-33-1 | MFCD00002136 | 98.0% | 158.09 g/mol | C6H7O3P | 100 G
SDP

Catalog No. 5000208551
Encompass_Preferred

Phenylphosphonic acid is an organophosphorus reagent used in chemical research and synthesis. It can serve as a phosphonic acid building block and as a ligand in certain metal-catalyzed reactions. Supplied as a white to off-white solid, it is commonly used in organic synthesis, materials research, and pesticide-related intermediate studies.

  • Phenylphosphonic acid, CAS 1571-33-1.
  • Purity 98.0%.
  • Molecular formula C6H7O3P, molecular weight 158.09 g/mol.
  • Appearance: solid, white to off-white.
  • Available in 100 G packages and other laboratory sizes.

Catalog No. 50-002-08551 Supplier Medchemexpress LLC Supplier No. HYW087962100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenylphosphonic acid is an organophosphorus reagent used in chemical research and synthesis. It can serve as a phosphonic acid building block and as a ligand in certain metal-catalyzed reactions. Supplied as a white to off-white solid, it is commonly used in organic synthesis, materials research, and pesticide-related intermediate studies.

  • Phenylphosphonic acid, CAS 1571-33-1.
  • Purity 98.0%.
  • Molecular formula C6H7O3P, molecular weight 158.09 g/mol.
  • Appearance: solid, white to off-white.
  • Available in 100 G packages and other laboratory sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.