Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenylphosphonic acid | 1571-33-1 | MFCD00002136 | 98.0% | 158.09 g/mol | C6H7O3P | 500 G
SDP

Catalog No. 5000208420
Encompass_Preferred

Phenylphosphonic acid is an organophosphorus reagent used as a chemical intermediate and building block in organic synthesis and research applications. It is supplied as a solid of 98.0% purity with molecular formula C6H7O3P and molecular weight 158.09 g/mol.

  • Organophosphorus reagent for organic synthesis and building-block chemistry.
  • Purity 98.0% suitable for research use.
  • Molecular formula C6H7O3P; molecular weight 158.09 g/mol.
  • Available in multiple pack sizes, including 500 G.

Catalog No. 50-002-08420 Supplier Medchemexpress LLC Supplier No. HYW087962500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenylphosphonic acid is an organophosphorus reagent used as a chemical intermediate and building block in organic synthesis and research applications. It is supplied as a solid of 98.0% purity with molecular formula C6H7O3P and molecular weight 158.09 g/mol.

  • Organophosphorus reagent for organic synthesis and building-block chemistry.
  • Purity 98.0% suitable for research use.
  • Molecular formula C6H7O3P; molecular weight 158.09 g/mol.
  • Available in multiple pack sizes, including 500 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.