Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenylphosphonic Dichloride

Catalog No. 389560100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-956-0100ML 100 mL
38-956-0500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-956-0100ML Supplier Sigma Aldrich Supplier No. 389560100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phenylphosphonic Dichloride

Specifications

CAS 824-72-6
Melting Point 3°C
Density 1.375 g/mL at 25°C
Boiling Point 258°C
Molecular Formula C6H5Cl2OP
Linear Formula C6H5POCl2
Refractive Index n20/D 1.559
MDL Number MFCD00002070
Quantity 100 mL
Synonym Benzenephosphonic dichloride; Benzenephosphonyl chloride; Phenyldichlorophosphine oxide; Phenylphosphonyl dichloride; Phenylphosphoryl dichloride; P,P-Dichlorophenylphosphine oxide
Molecular Weight (g/mol) 194.98 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.