Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phenylphosphonic dichloride, 97%

Catalog No. AAH3603714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3603714 25 g
AAH3603722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3603714 Supplier Thermo Scientific™ Supplier No. H3603714
Add to Cart

CAS: 824-72-6 | C6H5Cl2OP | 194.98 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-72-6
Molecular Formula C6H5Cl2OP
Molecular Weight (g/mol) 194.98
MDL Number MFCD00002070
InChI Key IBDMRHDXAQZJAP-UHFFFAOYSA-N
Synonym P,P-Dichlorophenylphosphine oxide;
IUPAC Name phenylphosphonoyl dichloride
SMILES ClP(Cl)(=O)C1=CC=CC=C1

Specifications

Melting Point 1-3°C
Density 1.379
Boiling Point 258°
Flash Point 204°(399°F)
Quantity 25 g
UN Number UN3265
Formula Weight 194.99
Percent Purity 97%
Physical Form Liquid
ShelfLife 5 Years
EINECSNumber 212-534-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.