Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylphosphonic Dichloride 98.0+%, TCI America™
SDP

Catalog No. P0334500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0334500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0334500ML Supplier TCI America Supplier No. P0334500ML

Specifications

CAS 824-72-6
Color Yellow
Boiling Point 258°C
Molecular Formula C6H5Cl2OP
Quantity 500 mL
MDL Number MFCD00002070
UN Number 3265
Synonym phenylphosphonic dichloride, phosphonic dichloride, phenyl, benzenephosphonic dichloride, phenylphosphonyl dichloride, dichlorophenylphosphine oxide, phenyldichlorophosphine oxide, benzenephosphonyl chloride, phenylphosphonic acid dichloride, phenylphosphonodichloridic acid, phosphonic dichloride, p-phenyl
InChI Key IBDMRHDXAQZJAP-UHFFFAOYSA-N
SMILES ClP(Cl)(=O)C1=CC=CC=C1
IUPAC Name phenylphosphonoyl dichloride
Molecular Weight (g/mol) 194.98
PubChem CID 69990
Formula Weight 194.98
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Phenylphosphonic Dichloride
Show More Show Less
EINECSNumber (3)-2611
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.