Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich phenylphosphoric acid

Catalog No. CDS0105461G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS0105461G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS0105461G Supplier Sigma Aldrich Supplier No. CDS0105461G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

cis-1,3-Dimethylcyclohexane undergoes ring opening reaction via dicarbene mechanism in the presence of iridium catalyst supported on SiO2.1,3-Dimethylcyclohexane was used to study ring opening reaction of 1,3-dimethylcyclohexane over Iridium catalysts modified by the addition of K and Ni.

Specifications

Molecular Formula C6H7O4P
Linear Formula C6H7O4P
MDL Number MFCD00058970
Quantity 25 g
Molecular Weight (g/mol) 174.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.