Learn More
Cayman Chemical PhenylpIperazInhydrochLRI 50mg
SDP
Catalog No. 502951188
Encompass_Preferred
The base compound from which bioactive drugs, such as the entactogen mCPP, are derived; intended for forensic applications
Catalog No. 50-295-1188 Supplier Cayman Chemical Supplier No. 1120350 MG
The base compound from which bioactive drugs, such as the entactogen mCPP, are derived; intended for forensic applications