Learn More
Cayman Chemical Phenylpiperazine (hydrochloride), Purity: ≥98%, CAS: 4004-95-9, 100 mg
SDP
Catalog No. 502951531
Encompass_Preferred
The base compound from which bioactive drugs, such as the entactogen mCPP, are derived; intended for forensic applications
Catalog No. 50-295-1531 Supplier Cayman Chemical Supplier No. 11203100 MG
The base compound from which bioactive drugs, such as the entactogen mCPP, are derived; intended for forensic applications