Learn More
Medchemexpress LLC Phenylpropiolic acid | 637-44-5 | 146.14 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0073195G
Encompass_Preferred
Phenylpropiolic acid is an endogenous metabolite and also functions as a click chemistry reagent. It contains an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Functions as an endogenous metabolite
  • Acts as a click chemistry reagent
  • Contains an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-004-60267