Learn More
Medchemexpress LLC Phenylpyruvic acid | 156-06-9 | 93.3% | 164.16 | 50 MG
SDP
Catalog No. 5000453687
Encompass_Preferred
Phenylpyruvic acid is an analytical standard intended for research and analytical applications. It acts as a precursor for the antifungal compound phenyllactic acid and can enhance the antifungal activity of eight lactic acid bacterial strains. It also demonstrates improved inhibitory activity against fungal bread contaminants such as Aspergillus niger and Penicillium roqueforti. This compound is commonly used as a reference standard in qualitative, quantitative, and methodological research experiments involving techniques like HPLC, GC, and MS.
Catalog No. 50-004-53687 Supplier Medchemexpress LLC Supplier No. HYW012530R50MG
Phenylpyruvic acid is an analytical standard intended for research and analytical applications. It acts as a precursor for the antifungal compound phenyllactic acid and can enhance the antifungal activity of eight lactic acid bacterial strains. It also demonstrates improved inhibitory activity against fungal bread contaminants such as Aspergillus niger and Penicillium roqueforti. This compound is commonly used as a reference standard in qualitative, quantitative, and methodological research experiments involving techniques like HPLC, GC, and MS.