Learn More
Medchemexpress LLC Phenylpyruvic acid | 156-06-9 | 99.9% | 164.16 | 500 MG
SDP
Catalog No. 5000455085
Encompass_Preferred
Phenylpyruvic acid is a precursor of the antifungal compound phenyllactic acid. It can improve the antifungal activity of eight lactic acid bacterial strains when added to a defined growth medium and demonstrates improved inhibitory activity against fungal bread contaminants. Additionally, it affects the enzyme activity of the pentose phosphate pathway involved in the oxidative phase in rat brain homogenates and can reduce glucose-6-phosphate dehydrogenase activity.
  • Increases inhibitory activity against fungal contaminants from 13.5% to 46.5%.
  • Increases phenyllactic acid production up to six-fold for lactic acid bacterial strains.
  • Significantly reduces glucose-6-phosphate dehydrogenase (G6PD) activity at 0.6 and 1.2 mM without pre-incubation.
Catalog No. 50-004-55085 Supplier Medchemexpress LLC Supplier No. HYW012530500MG
Phenylpyruvic acid is a precursor of the antifungal compound phenyllactic acid. It can improve the antifungal activity of eight lactic acid bacterial strains when added to a defined growth medium and demonstrates improved inhibitory activity against fungal bread contaminants. Additionally, it affects the enzyme activity of the pentose phosphate pathway involved in the oxidative phase in rat brain homogenates and can reduce glucose-6-phosphate dehydrogenase activity.
  • Increases inhibitory activity against fungal contaminants from 13.5% to 46.5%.
  • Increases phenyllactic acid production up to six-fold for lactic acid bacterial strains.
  • Significantly reduces glucose-6-phosphate dehydrogenase (G6PD) activity at 0.6 and 1.2 mM without pre-incubation.