Learn More
Medchemexpress LLC Phenylpyruvic acid | 156-06-9 | MFCD00002589 | 99.9% | 164.16 | 25MG
SDP
Catalog No. 5000452677
Encompass_Preferred
Phenylpyruvic acid is a precursor of the antifungal compound phenyllactic acid. It affects the enzyme activity of the pentose phosphate pathway involved in the oxidative phase in rat brain homogenates and can reduce glucose-6-phosphate dehydrogenase activity. For research use only; not sold to patients.
  • Can improve the antifungal activity of eight lactic acid bacterial strains when added to a defined growth medium.
  • Demonstrates improved inhibitory activity against fungal bread contaminants such as *Aspergillus niger* and *Penicillium roqueforti*.
Catalog No. 50-004-52677 Supplier Medchemexpress LLC Supplier No. 25MGHYW012530R
Phenylpyruvic acid is a precursor of the antifungal compound phenyllactic acid. It affects the enzyme activity of the pentose phosphate pathway involved in the oxidative phase in rat brain homogenates and can reduce glucose-6-phosphate dehydrogenase activity. For research use only; not sold to patients.
  • Can improve the antifungal activity of eight lactic acid bacterial strains when added to a defined growth medium.
  • Demonstrates improved inhibitory activity against fungal bread contaminants such as *Aspergillus niger* and *Penicillium roqueforti*.