Learn More
Cayman Chemical Phenylpyruvic Acid, Purity: ≥95%, CAS: 156-06-9, 5 g
SDP
Catalog No. 502912208
Encompass_Preferred
An active metabolite of phenylalanine; a biosynthetic precursor to 3-phenyllactic acid; inhibits G6PDH in rat brain homogenates at 0.6 and 1.2 mM; increases 3-phenyllactic acid levels in, and the antifungal activity of, the cell-free culture supernatant from lactic acid bacteria; urinary levels are increased in patients with PKU
Catalog No. 50-291-2208 Supplier Cayman Chemical Supplier No. 363635 G
An active metabolite of phenylalanine; a biosynthetic precursor to 3-phenyllactic acid; inhibits G6PDH in rat brain homogenates at 0.6 and 1.2 mM; increases 3-phenyllactic acid levels in, and the antifungal activity of, the cell-free culture supernatant from lactic acid bacteria; urinary levels are increased in patients with PKU