Learn More
Medchemexpress LLC Phenylpyruvic acid (sodium) | 114-76-1 | MFCD00002590 | 98.2% | 186.14 g/mol | C9H7NaO3 | 25 MG
SDP
Catalog No. 5000304043
Encompass_Preferred
Phenylpyruvic acid (sodium) is the sodium salt form of phenylpyruvic acid provided for research use as a high-purity biochemical reagent (CAS 114-76-1). It is a white to off-white solid (C9H7NaO3, MW 186.14 g/mol) commonly used in metabolic studies and enzymatic assays and is supplied in laboratory-scale quantities.
  • High purity (~98.2%) suitable for analytical and biochemical applications.
  • Water-soluble sodium salt form for convenient preparation of aqueous solutions.
  • Appropriate for metabolic and enzymatic studies as a biochemical substrate.
  • Available in small lab-friendly package sizes for experimental use.
  • Stable solid form for straightforward storage and handling.
Catalog No. 50-003-04043 Supplier Medchemexpress LLC Supplier No. 25MGHYW016203
Phenylpyruvic acid (sodium) is the sodium salt form of phenylpyruvic acid provided for research use as a high-purity biochemical reagent (CAS 114-76-1). It is a white to off-white solid (C9H7NaO3, MW 186.14 g/mol) commonly used in metabolic studies and enzymatic assays and is supplied in laboratory-scale quantities.
  • High purity (~98.2%) suitable for analytical and biochemical applications.
  • Water-soluble sodium salt form for convenient preparation of aqueous solutions.
  • Appropriate for metabolic and enzymatic studies as a biochemical substrate.
  • Available in small lab-friendly package sizes for experimental use.
  • Stable solid form for straightforward storage and handling.