Learn More
Medchemexpress LLC Phenylpyruvic acid sodium | 114-76-1 | MFCD00002590 | 98.2% | 186.14 g/mol | C9H7NaO3 | 250 MG
SDP
Catalog No. 5000302574
Encompass_Preferred
Phenylpyruvic acid sodium is the sodium salt of phenylpyruvic acid used as a research reagent. It is provided as a high-purity solid with supporting analytical and safety documentation, making it suitable for biochemical investigations and synthetic chemistry applications.
  • High purity (98.2%) for reliable experimental results.
  • Molecular weight 186.14 g/mol and formula C9H7NaO3.
  • Supplied as a dry solid, convenient for weighing and storage.
  • Includes certificate of analysis and safety data sheet for quality and handling information.
  • Store sealed away from moisture; recommended solvent storage -80°C for 6 months or -20°C for 1 month.
  • Suitable for biochemical research and synthetic applications.
Catalog No. 50-003-02574 Supplier Medchemexpress LLC Supplier No. 250MGHYW016203
Phenylpyruvic acid sodium is the sodium salt of phenylpyruvic acid used as a research reagent. It is provided as a high-purity solid with supporting analytical and safety documentation, making it suitable for biochemical investigations and synthetic chemistry applications.
  • High purity (98.2%) for reliable experimental results.
  • Molecular weight 186.14 g/mol and formula C9H7NaO3.
  • Supplied as a dry solid, convenient for weighing and storage.
  • Includes certificate of analysis and safety data sheet for quality and handling information.
  • Store sealed away from moisture; recommended solvent storage -80°C for 6 months or -20°C for 1 month.
  • Suitable for biochemical research and synthetic applications.