Learn More
Medchemexpress LLC Phenylpyruvic acid (sodium) | 114-76-1 | MFCD00002590 | 98.2% | 186.14 g/mol | C9H7NaO3 | 500 MG
SDP
Catalog No. 5000303693
Encompass_Preferred
Phenylpyruvic acid sodium is the sodium salt of phenylpyruvic acid supplied as a crystalline reagent for laboratory and biochemical research. It is used as an endogenous metabolite standard and a reagent in studies of phenylalanine metabolism and enzyme assays.
  • High purity suitable for analytical and preparative work.
  • Known molecular weight 186.14 g/mol and formula C9H7NaO3.
  • Documented identity with CAS 114-76-1 for traceability.
  • Provided with SDS and COA documentation for safe handling and quality verification.
  • Available in multiple common laboratory sizes for flexibility.
Catalog No. 50-003-03693 Supplier Medchemexpress LLC Supplier No. HYW016203500MG
Phenylpyruvic acid sodium is the sodium salt of phenylpyruvic acid supplied as a crystalline reagent for laboratory and biochemical research. It is used as an endogenous metabolite standard and a reagent in studies of phenylalanine metabolism and enzyme assays.
  • High purity suitable for analytical and preparative work.
  • Known molecular weight 186.14 g/mol and formula C9H7NaO3.
  • Documented identity with CAS 114-76-1 for traceability.
  • Provided with SDS and COA documentation for safe handling and quality verification.
  • Available in multiple common laboratory sizes for flexibility.