Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenylselenyl bromide

Catalog No. 24396525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-396-525G 25 g
24-396-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-396-525G Supplier Sigma Aldrich Supplier No. 24396525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phenylselenyl bromide

Specifications

CAS 34837-55-3
Melting Point 58°C to 62°C (lit.)
Boiling Point 107°C to 108°C (15 mmHg)
Molecular Formula C6H5BrSe
Linear Formula C6H5SeBr
MDL Number MFCD00000047
Quantity 25 g
Synonym Benzeneselenenyl bromide; Benzeneselenyl bromide
Molecular Weight (g/mol) 235.97
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.