Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenylselenyl chloride

Catalog No. 18334210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-334-210G 10 g
18-334-21G 1 g
18-334-250G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-334-210G Supplier Sigma Aldrich Supplier No. 18334210G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phenylselenyl chloride

Specifications

CAS 5707-04-0
Melting Point 59°C to 62°C (lit.)
Boiling Point 120°C (20 mmHg)
Molecular Formula C6H5ClSe
Linear Formula C6H5SeCl
MDL Number MFCD00000478
Quantity 10 g
Synonym Benzeneselenenyl chloride; Benzeneselenyl chloride
Molecular Weight (g/mol) 191.52
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.