Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenylsilane

Catalog No. 3351505G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-515-05G 5 g
33-515-025G 25 g
33-515-01G 1 g
33-515-0500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 33-515-05G Supplier Sigma Aldrich Supplier No. 3351505G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phenylsilane (PhSiH3) may be used as a reducing agent for the partial reduction of phosphine oxide groups in poly(4,4'sa-diphenylphenylphosphine oxide) (PAPO) to phosphine. It can also undergo photopolymerization with methyl methacrylate(MMA) to form poly(MMA) containing SiH groups. PhSiH3 may be used as a silicon source to synthesize crystalline Si nanowires via Au-nanocrystal-seeded supercritical fluid-liquid-solid (SFLS) synthesis.

Specifications

CAS 694-53-1
Density 0.877 g/mL at 25°C
Boiling Point 120°C
Molecular Formula C6H8Si
Refractive Index n20/D 1.510
Linear Formula C6H5SiH3
MDL Number MFCD00013478
Quantity 5 g
Molecular Weight (g/mol) 108.21 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.