Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylsulfur Pentafluoride 98.0+%, TCI America™
SDP

Catalog No. P19691G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19691G Supplier TCI America Supplier No. P19691G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2557-81-5
Molecular Formula C6H5F5S
Molecular Weight (g/mol) 204.158
MDL Number MFCD09263740
InChI Key DMNYFEWFSFTYEL-UHFFFAOYSA-N
PubChem CID 9942464
IUPAC Name pentafluoro(phenyl)-$l^{6}-sulfane
SMILES C1=CC=C(C=C1)S(F)(F)(F)(F)F

Specifications

Melting Point -10°C
Color Colorless
Boiling Point 149°C
Quantity 1 g
UN Number 1993
Formula Weight 204.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenylsulfur Pentafluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.