Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Phenylthio)trimethylsilane 95.0+%, TCI America™
SDP

Catalog No. P13785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13785G 5 g
P137825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13785G Supplier TCI America Supplier No. P13785G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4551-15-9
Molecular Formula C9H14SSi
Molecular Weight (g/mol) 182.356
MDL Number MFCD00008272
InChI Key VJMQFIRIMMSSRW-UHFFFAOYSA-N
Synonym phenylthiotrimethylsilane, phenylthio trimethylsilane, silane, trimethyl phenylthio, phenyl trimethylsilyl sulfide, trimethyl phenylthio silane, trimethyl phenylsulfanyl silane, benzene, trimethylsilyl thio, s-trimethylsilylthiophenol, phenylthio trimethyl silane
PubChem CID 78312
IUPAC Name trimethyl(phenylsulfanyl)silane
SMILES C[Si](C)(C)SC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 65°C
Quantity 5 g
UN Number 1993
Formula Weight 182.36
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (Phenylthio)trimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.