Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylthiohydantoin-DELTA-threonine 90.0+%, TCI America™
SDP

Catalog No. P03701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03701G 1 g
P0370100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P03701G Supplier TCI America Supplier No. P03701G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5800-50-0
Molecular Formula C11H10N2OS
Molecular Weight (g/mol) 218.274
MDL Number MFCD00070617
InChI Key SXYSWGDIVRMEDE-MBXJOHMKSA-N
Synonym 5-Ethylidene-3-phenyl-2-thiohydantoin, PTH-DELTA-threonine
PubChem CID 45105912
IUPAC Name (5Z)-5-ethylidene-3-phenyl-2-sulfanylideneimidazolidin-4-one
SMILES CC=C1C(=O)N(C(=S)N1)C2=CC=CC=C2

Specifications

Melting Point 236°C
Quantity 1 g
Formula Weight 218.27
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenylthiohydantoin-DELTA-threonine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.