Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylthiohydantoin-tyrosine 98.0+%, TCI America™
SDP

Catalog No. P0368100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0368100MG 100 mg
P03681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0368100MG Supplier TCI America Supplier No. P0368100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4332-95-0
Molecular Formula C16H14N2O2S
Molecular Weight (g/mol) 298.36
MDL Number MFCD00027415
InChI Key LCFDDJINTNHAEQ-UHFFFAOYSA-N
Synonym 5-(4-Hydroxybenzyl)-3-phenyl-2-thiohydantoin, PTH-tyrosine
PubChem CID 2872399
IUPAC Name 5-[(4-hydroxyphenyl)methyl]-3-phenyl-2-sulfanylideneimidazolidin-4-one
SMILES C1=CC=C(C=C1)N2C(=O)C(NC2=S)CC3=CC=C(C=C3)O

Specifications

Melting Point 221°C
Color White-Yellow
Quantity 100 mg
Formula Weight 298.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenylthiohydantoin-tyrosine
RTECSNumber NJ0730000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.