Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylthiohydantoin-valine 98.0+%, TCI America™
SDP

Catalog No. P0379100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0379100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0379100MG Supplier TCI America Supplier No. P0379100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4333-20-4
Molecular Formula C12H14N2OS
Molecular Weight (g/mol) 234.32
MDL Number MFCD00038438
InChI Key HEIKHJHSVLQGGN-UHFFFAOYNA-N
Synonym 5-Isopropyl-3-phenyl-2-thiohydantoin, PTH-valine
PubChem CID 2831422
IUPAC Name 3-phenyl-5-(propan-2-yl)-2-sulfanylideneimidazolidin-4-one
SMILES CC(C)C1NC(=S)N(C1=O)C1=CC=CC=C1

Specifications

Melting Point 209°C
Color White
Quantity 100 mg
Formula Weight 234.32
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenylthiohydantoin-valine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.