Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Phenylthiomethyl)trimethylsilane 98.0+%, TCI America™
SDP

Catalog No. P11725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11725ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11725ML Supplier TCI America Supplier No. P11725ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17873-08-4
Molecular Formula C10H16SSi
Molecular Weight (g/mol) 196.383
MDL Number MFCD00009838
InChI Key UOQDIMYVQSHALM-UHFFFAOYSA-N
Synonym Phenylthio(trimethylsilyl)methane, Phenyl Trimethylsilylmethyl Sulfide, Trimethyl(phenylthiomethyl)silane
PubChem CID 140295
IUPAC Name trimethyl(phenylsulfanylmethyl)silane
SMILES C[Si](C)(C)CSC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 158°C
Quantity 5 mL
UN Number 1993
Formula Weight 196.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Phenylthiomethyl)trimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.