Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylthiophosphonic Dichloride 98.0+%, TCI America™
SDP

Catalog No. P1245500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1245500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1245500G Supplier TCI America Supplier No. P1245500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3497-00-5
Molecular Formula C6H5Cl2PS
Molecular Weight (g/mol) 211.04
MDL Number MFCD00004916
InChI Key SXIWNIQDOJKDGB-UHFFFAOYSA-N
Synonym Phenylphosphonothioic Dichloride
PubChem CID 19016
IUPAC Name dichloro-phenyl-sulfanylidene-$l^{5}-phosphane
SMILES C1=CC=C(C=C1)P(=S)(Cl)Cl

Specifications

Color Yellow
Boiling Point 270°C
Quantity 500 g
UN Number 3265
Formula Weight 211.04
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Phenylthiophosphonic Dichloride
EINECSNumber (3)-2511&(3)-2612
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.