Learn More
Medchemexpress LLC Phenylthiourea | 103-85-5 | MFCD00004933 | 99.3% | 152.22 g/mol | C7H8N2S | 100g
SDP
Catalog No. 5000269689
Encompass_Preferred
Phenylthiourea (Phenylthiocarbamide) is an inhibitor for phenoloxidase Phenylthiourea inhibits enzymatic oxidation of DOPA by phenoloxidase (Ki 0 21 M) Phenylthiourea is an effective inhibitor for tyrosinase Phenylthiourea can lead to graying of hair in black rats due to the interference with melanin formation[1][2]
Catalog No. 50-002-69689 Supplier Medchemexpress LLC Supplier No. HYY0351100G
Phenylthiourea (Phenylthiocarbamide) is an inhibitor for phenoloxidase Phenylthiourea inhibits enzymatic oxidation of DOPA by phenoloxidase (Ki 0 21 M) Phenylthiourea is an effective inhibitor for tyrosinase Phenylthiourea can lead to graying of hair in black rats due to the interference with melanin formation[1][2]