Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenyltin Trichloride

Catalog No. 2772315G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-723-15G 5 g
27-723-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 27-723-15G Supplier Sigma Aldrich Supplier No. 2772315G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phenyltin trichloride can be used:In the preparation of organotin(IV) complexes as potent antimicrobial agents.As a reactant in the Stille coupling reaction catalyzed by peptide-templated Pd nanoparticles.As a tin (Sn) source in the synthesis of tin-doped TiO2 photoanodes applicable in dye-sensitized solar cells.

Specifications

CAS 1124-19-2
Density 1.839 g/mL at 25°C
Boiling Point 142°C to 143°C (25 mmHg, lit.)
Molecular Formula C6H5Cl3Sn
Refractive Index n20/D 1.585
Linear Formula C6H5SnCl3
MDL Number MFCD00000513
Quantity 5 g
Synonym Trichlorophenylstannane
Molecular Weight (g/mol) 302.17 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.