Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyltrichlorosilane 98.0+%, TCI America™
SDP

Catalog No. P0240500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0240500G 500 g
P024025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0240500G Supplier TCI America Supplier No. P0240500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 98-13-5
Molecular Formula C6H5Cl3Si
Molecular Weight (g/mol) 211.541
MDL Number MFCD00000479
InChI Key ORVMIVQULIKXCP-UHFFFAOYSA-N
Synonym trichloro phenyl silane, phenyltrichlorosilane, silane, trichlorophenyl, phenyl trichlorosilane, phenylsilicon trichloride, silane, phenyltrichloro, silicon phenyl trichloride, benzene, trichlorosilyl, unii-m7199jl06m, phenyl-trichloro-silane
PubChem CID 7372
IUPAC Name trichloro(phenyl)silane
SMILES C1=CC=C(C=C1)[Si](Cl)(Cl)Cl

Specifications

Color Colorless
Boiling Point 201°C
Quantity 500 g
UN Number 1804
Formula Weight 211.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyltrichlorosilane
EINECSNumber (3)-2634
RTECSNumber VV6650000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.