Learn More
Medchemexpress LLC Phenyltrimethylammonium chloride | 138-24-9 | MFCD00011790 | ≥98.0% | 171.67 g/mol | C9H14ClN | 25 G
SDP
Catalog No. 5000302319
Encompass_Preferred
Phenyltrimethylammonium chloride (CAS 138-24-9) is a quaternary ammonium chloride used as a phase-transfer catalyst and organic reagent in synthesis. It is supplied as a high-purity chloride salt for laboratory research and small-scale synthesis applications.
  • High purity (≥98.0%) suitable for research-grade reactions.
  • Functions as a phase-transfer catalyst to facilitate biphasic reactions.
  • Stable, easy-to-handle solid for accurate weighing and dosing.
  • Soluble in common organic solvents for versatile reaction conditions.
  • Available in gram-scale packs to support method development and scale-up.
Catalog No. 50-003-02319 Supplier Medchemexpress LLC Supplier No. 25GHYW016923
Phenyltrimethylammonium chloride (CAS 138-24-9) is a quaternary ammonium chloride used as a phase-transfer catalyst and organic reagent in synthesis. It is supplied as a high-purity chloride salt for laboratory research and small-scale synthesis applications.
  • High purity (≥98.0%) suitable for research-grade reactions.
  • Functions as a phase-transfer catalyst to facilitate biphasic reactions.
  • Stable, easy-to-handle solid for accurate weighing and dosing.
  • Soluble in common organic solvents for versatile reaction conditions.
  • Available in gram-scale packs to support method development and scale-up.