Learn More
Medchemexpress LLC Phenyltrimethylammonium chloride | 138-24-9 | MFCD00011790 | ≥98.0% | 171.67 g/mol | C9H14ClN | 500 G
SDP
Catalog No. 5000299878
Encompass_Preferred
N,N,N-Trimethylbenzenaminium chloride is a quaternary ammonium salt used as a reagent in organic synthesis and as a phase-transfer catalyst. It is supplied as a solid (pale purple to purple) with a defined chemical formula and molecular weight for laboratory use.
  • High purity: ≥98.0%.
  • Functions as a phase-transfer catalyst to facilitate biphasic reactions.
  • Useful reagent for methylation and other organic transformations.
  • Solid form with consistent physical appearance for ease of handling.
  • Well-characterized chemical identity (CAS 138-24-9) for reproducibility.
Catalog No. 50-002-99878 Supplier Medchemexpress LLC Supplier No. HYW016923500G
N,N,N-Trimethylbenzenaminium chloride is a quaternary ammonium salt used as a reagent in organic synthesis and as a phase-transfer catalyst. It is supplied as a solid (pale purple to purple) with a defined chemical formula and molecular weight for laboratory use.
  • High purity: ≥98.0%.
  • Functions as a phase-transfer catalyst to facilitate biphasic reactions.
  • Useful reagent for methylation and other organic transformations.
  • Solid form with consistent physical appearance for ease of handling.
  • Well-characterized chemical identity (CAS 138-24-9) for reproducibility.