Learn More
U.S. Pharmacopeia Phenytoin Related Compound B, 6802-95-5, MFCD01733136, 50 mg
Catalog No. NC1403959
Encompass
Empirical Formula (Hill Notation): C15H14N2O3, Molecular Weight: 270.28, Synonym: α-[(Aminocarbonyl)amino]-α-phenylbenzeneacetic acid.
Catalog No. NC1403959 Supplier U.S. Pharmacopeia Supplier No. 1535020
Empirical Formula (Hill Notation): C15H14N2O3, Molecular Weight: 270.28, Synonym: α-[(Aminocarbonyl)amino]-α-phenylbenzeneacetic acid.