Learn More
Medchemexpress LLC Phenytoin-15N2,13C | 78213-26-0 | 99.8% | 120 UG
SDP
Catalog No. 5000407824
Encompass_Preferred
Phenytoin-15N2,13C is a 13C and 15N labeled derivative of Phenytoin (5,5-Diphenylhydantoin), known for its potent action as a Voltage-gated Na+ channels (VGSCs) blocker. It demonstrates antiepileptic activity and has been observed to reduce breast tumor growth and metastasis in mice. This compound incorporates stable heavy isotopes, making it suitable for use as a tracer in drug development for quantitative analysis.
  • Can be utilized as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits antiepileptic activity
  • Reduces breast tumor growth and metastasis
  • Potential to influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-07824 Supplier Medchemexpress LLC Supplier No. HYB0448S1120UG
Phenytoin-15N2,13C is a 13C and 15N labeled derivative of Phenytoin (5,5-Diphenylhydantoin), known for its potent action as a Voltage-gated Na+ channels (VGSCs) blocker. It demonstrates antiepileptic activity and has been observed to reduce breast tumor growth and metastasis in mice. This compound incorporates stable heavy isotopes, making it suitable for use as a tracer in drug development for quantitative analysis.
  • Can be utilized as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits antiepileptic activity
  • Reduces breast tumor growth and metastasis
  • Potential to influence pharmacokinetic and metabolic profiles of drugs